


Yttrium (III) nitrate hexahydrate, Sigma-Aldrich®
Catalog # 1.12516.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:1500 g/l (20 °C)
- Molar Mass:383.01 g/mol
- Bulk Density:1100 kg/m3
- Density:1.1 g/cm3 (20 °C)
- pH:3.5 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
About this item
:
:383.01 g/mol
Melting Pt:51,8 °C
Density:2,682 g/cm³ (20 °C)
ADR:5.1,
:M-Clarity™ quality level = MQ200
EINECS:233-802-6
UN:1477
:GHS03
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:13494-98-9
Formula:N₃O₉Y·6H₂O
MDL Number:MFCD00149940