


Zinc sulphate heptahydrate 99% ACS
:7446-20-0
:
:ZnSO₄·7H₂O
:MFCD00149894
:12,10293
:100 °C
:1,97 g/cm³ (20 °C)
:Ambient
:231-793-3
:3077
:9,III
:287.54 g/mol
:GHS07
:GHS05
:GHS09
- Appearance:White Crystals and/or chunks
- Titration Complexometric:99.0 to 103.0 %
- pH:4.4 to 6.0 (5 % at 25°C)
- Chloride (Cl):≤5 ppm
- Nitrate (NO3):≤20 ppm
- Ammonium (NH4):≤10 ppm
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤10 ppm
- Lead (Pb):≤30 ppm
- Magnesium (Mg):≤50 ppm
- Manganese (Mn):≤3 ppm
- Potassium (K):≤100 ppm
- Sodium (Na):≤500 ppm
- Insoluble matter:≤0.01 %
Frequently Bought Together