Copper(II) nitrate hemipentahydrate ≥97%, TECHNICAL
: Cupric Nitrate Hydrate, Copper dinitrate hemipentahydrate, Cupric nitrate hemi(pentahydrate), Cupric nitrate 2.5-hydrate, Cupric nitrate hemipentahydrate
:19004-19-4
:
:Cu(NO₃)₂·2.5H₂O
:MFCD02253001
:232.59 g/mol
:114 °C
:2,05 g/cm³ (20 °C)
:221-838-5
:1477
:5.1,II
:GHS03
:GHS07
:GHS09
- Environmentally Preferable:
- Assay:Min. 97 %
Frequently Bought Together