Order Entry
Northern Ireland
ContactUsLinkComponent
Copper(II) chloride dihydrate ≥99.0% ACS, Fluka™
Environment
Irritant
Corrosive
Copper(II) chloride dihydrate ≥99.0% ACS, Fluka™
Catalog # 307483-100G
CAS Number:  
Environment
Irritant
Corrosive
Copper(II) chloride dihydrate ≥99.0% ACS, Fluka™
Catalog # 307483-100G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • pH (5 %, 20°C):
    3.0 - 3.8
  • Insoluble in hydrochloric acid:
    Max. 0.01 %
  • Calcium (Ca):
    Max. 50 ppm
  • Iron (Fe):
    Max. 50 ppm
  • Potassium (K):
    Max. 100 ppm
  • Sodium (Na):
    Max. 200 ppm
  • Nickel (Ni):
    Max. 100 ppm
  • Nitrate (NO₃):
    Max. 0.015 %
  • Sulfate (SO₄):
    Max. 50 ppm

Specification Test Results

About this item

 :
 :170.48 g/mol
Merck Index:13,02660
Boiling Pt:993 °C (anhydr.)
Melting Pt:598 °C (anhy) 110 °C (–H₂O of crystall.)
Density:2,53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-210-2
UN:2802
ADR:8,III
 :GHS09
 :GHS07
 :GHS05
CAS Number:10125-13-0
Formula:CuCl₂·2H₂O
MDL Number:MFCD00149674