Order Entry
Northern Ireland
ContactUsLinkComponent
Terbium(III) sulphate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Irritant
Terbium(III) sulphate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Catalog # 14596.09
CAS Number:  
Irritant
Terbium(III) sulphate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Catalog # 14596.09
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :782.16 g/mol
Melting Pt:360 °C (-8H₂O)
EINECS:237-213-5
 :GHS07
Storage Temperature:Ambient
CAS Number:13842-67-6
Formula:O₁₄S₃Tb₂·8H₂O
MDL Number:MFCD00149872