
Cerium(IV) sulphate tetrahydrate ≥98.0% analytical reagent
Catalog # 22604.153
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Assay:Min. 98.0 %
- Heavy metals (as Pb):Max. 50 ppm
- Insolubility in diluted H2SO4:Max. 50 ppm
- Cl (Chloride):Max. 10 ppm
- PO4 (Phosphate):Max. 100 ppm
- Fe (Iron):Max. 50 ppm
Specification Test Results
About this item
:
Merck Index:12,02036
Melting Pt:180…200 °C
Density:3,91 g/cm³ (20 °C)
ADR:8,III
:404.3 g/mol
EINECS:237-029-5
UN:3260
:GHS07
CAS Number:10294-42-5
Formula:Ce(SO₄)₂·4H₂O
MDL Number:MFCD00149427