Order Entry
Northern Ireland
ContactUsLinkComponent
Cerium(IV) sulphate tetrahydrate, EMSURE® for analysis, Supelco®
Corrosive
Environment
Cerium(IV) sulphate tetrahydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
Corrosive
Environment
Cerium(IV) sulphate tetrahydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    38 g/l (50 °C)
  • Melting Point:
    180 - 200 °C Elimination of water of crystallisation
  • Molar Mass:
    404.30 g/mol
  • Bulk Density:
    650 - 850 kg/m3
  • Density:
    5.02 g/cm3 (20 °C)
  • pH:
    1.6 (10 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together