

Erbium trinitrate pentahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10031-51-3
:
Formula:ErN₃O₉·5H₂O
MDL Number:MFCD00149691
:443.35 g/mol
Melting Pt:130 °C (-4H₂O)
Storage Temperature:Ambient
EINECS:233-436-7
UN:1477
ADR:5.1,III
:GHS03
:GHS07
Related Information
Frequently Bought Together