

Cerium(III) nitrate hydrate ≥99.998% (metals basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10108-73-3
:
Formula:Ce(NO₃)₃·xH₂O
MDL Number:MFCD00010926
:326.13 g/mol (anhy)
Merck Index:14,2000
Boiling Pt:200 °C (1013 hPa)
Melting Pt:39 °C
Density:2,377 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-297-2
UN:1477
ADR:5.1,III
:GHS03
:GHS07
Frequently Bought Together