
Ytterbium(III) chloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19423-87-1
:MFCD00149884
Formula:Cl₃Yb·H₂O
MDL Number:MFCD00149885
:297.41 g/mol
Merck Index:14,10105
Melting Pt:150…155 °C
Density:2,58
EINECS:233-800-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together