


Lanthanum(III) chloride hydrate ≥98.5% (by titration analysis on dried basis) (~7 mol/mol H₂O)
Catalog # 61490-500G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Lanthanum chloride (LaCl₃):64.5 - 70.0 %
- Water (Karl Fischer):32 - 36 %
- Calcium (Ca):Max. 50 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 10 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 5 ppm
- Sodium (Na):Max. 10 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Sulfate (SO₄):Max. 50 ppm
Specification Test Results
About this item
:
:263.28
Boiling Pt:? 860 °C (1013 hPa)
Melting Pt:92-93 °C
Melting Pt:92…93 °C
Storage Temperature:Ambient
EINECS:233-237-5
UN:3260
ADR:8,III
:GHS09
:GHS07
:GHS05
CAS Number:20211-76-1
:10025-84-0
Formula:Cl₃La·H₂O
MDL Number:MFCD00149753