
Gadolinium trichloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13450-84-5
:
Formula:Cl₃Gd·6H₂O
MDL Number:MFCD00149732
:371.7 g/mol
Merck Index:14,4326
Density:2,424
EINECS:233-386-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together