Europium(III) chloride hexahydrate 99.9% (trace metals basis) -10 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13759-92-7
:
Formula:Cl₃Eu·6H₂O
MDL Number:MFCD00149701
:366.41 g/mol
Melting Pt:850 °C
Density:4,89 g/cm³
EINECS:233-040-4
Specification Test Results
- Appearance:White Crystals
- Total conc. of imp.as REO:≤0.1 %
Specification Test Results
Frequently Bought Together