
Samarium tribromide hexahydrate 99%
Catalog # A18191.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:498.16 g/mol
Melting Pt:>640 °C
EINECS:237-347-4
:GHS07
Storage Temperature:Ambient
CAS Number:13517-12-9
Formula:Br₃Sm·6H₂O
MDL Number:MFCD00192430