



Tetramethylammonium hydroxide pentahydrate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10424-65-4
:
Formula:C₄H₁₃NO·5H₂O
MDL Number:MFCD00149566
:181.23 g/mol
Melting Pt:62…71 °C
Density:1,829 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-882-9
UN:3423
ADR:8,II
:GHS06
:GHS05
:GHS08
:GHS09
Specification Test Results
- Appearance:White to off-white Hygroscopic powder or crystals
- Infrared spectrum:Conforms
- Melting point:62 to 73 °C
- Titration with HCl:≥96.0 %
Specification Test Results
Frequently Bought Together