ContactUsLinkComponent
Bole white (kaolin), pure
undefined
Bole white (kaolin), pure
  • Appearance:
    Light beige Powder
  • Loss on drying:
    ≤1.5 %
  • Particle size:
    50 to 62 % (< 2 µm)
  • Loss on ignition:
    10.8 % to 11.8 %
Frequently Bought Together