Calcium sulphate dihydrate, EMSURE® for analysis, precipitated, Supelco®
: Merck
:10101-41-4
:
:CaSO₄·2H₂O
:MFCD00149625
:172.17 g/mol
:13,01711
:2,32 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:Ambient
:231-900-3
- Solubility:2 g/l (20 °C)
- Molar Mass:172.17 g/mol
- Bulk Density:400 - 600 kg/m3
- Density:2.32 g/cm3 (20 °C)
- pH:7.0 (50 g/l, H2O, 20 °C) (slurry)
Frequently Bought Together