
Strontium bromide hexahydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-53-9
:
Formula:Br₂Sr·6H₂O
MDL Number:MFCD00150361
:355.52 g/mol
Merck Index:14,8837
Density:2,386
EINECS:233-969-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together