Order Entry
Northern Ireland
ContactUsLinkComponent
Disodium sulphide nonahydrate 98+%, extra pure
Corrosive
Toxic
Environment
Disodium sulphide nonahydrate 98+%, extra pure
Catalog # ACRO387065000
CAS Number:  
undefined
Corrosive
Toxic
Environment
Disodium sulphide nonahydrate 98+%, extra pure
Catalog # ACRO387065000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    Colorless to light yellow Adhering crystals and/or chunks
  • Titration Iodimetric:
    32.0 to 38.0 % (Na2S)
  • Thiosulphate (S2O3):
    ≤5000 ppm
  • Sulfite (as SO2):
    ≤2000 ppm
  • Total nitrogen (as N):
    ≤0.005 %

Specification Test Results

About this item

 :
 :240.18 g/mol
Boiling Pt:920 °C (1013 hPa)
Melting Pt:50 °C
Density:1,427 g/cm³ (16 °C)
Storage Temperature:Refrigerator
EINECS:215-211-5
UN:1849
ADR:8,II
REACH:01-2119513694-38
 :GHS05
 :GHS06
 :GHS09
CAS Number:1313-84-4
Formula:Na₂S·9H₂O
MDL Number:MFCD00149184