

tetra-Chloroauric(III) acid trihydrate analytical reagent, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:16961-25-4
:
Formula:HAuCl₄·3H₂O
MDL Number:MFCD00149904
:393.83 g/mol
Melting Pt:30 °C
Density:3,9 g/cm³ (20 °C)
EINECS:240-948-4
UN:3260
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:150 g/l (20 °C)
- Melting Point:30 °C
- Molar Mass:393.83 g/mol
- Density:3.9 g/cm3 (20 °C)
- pH:(H2O) strongly acid
Specification Test Results
Related Information
Frequently Bought Together