trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 2,2',2'',2'''-(trans-Cyclohexane-1,2-diylbis(azanetriyl))tetraacetic acid hydrate, DCTA monohydrate, CDTA monohydrate, trans-Cyclohex-1,2-ylenediaminetetra acetic acid monohydrate, (1,2-Cyclohexylenedinitrilo)tetraacetic acid monohydrate, 2,2',2'',2'''-[trans-Cyclohexane-1,2-diylbis(azanetriyl)]tetraacetic acid hydrate, CHELATON 4, Titriplex® IV, trans-1,2-Cyclohexanediaminetetraacetic acid monohydrate, trans-Cyclohexane-1,2-dinitrilotetraacetic acid monohydrate
CAS Number:125572-95-4
:MFCD00066429
Formula:C₁₄H₂₂N₂O₈·1H₂O
Molecular Weight:364.35 g/mol
Melting Pt:213…216 °C
:M-Clarity™ quality level = MQ200
EINECS:236-308-9
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:213 °C
- Molar Mass:364.34 g/mol
- Bulk Density:500 kg/m3
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_841032
SDS
ProductPage_841032 ProductPage_841032
Product specification
Frequently Bought Together