Oxalic acid dihydrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0187.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:99.5 - 102.5 %
- Insoluble Matter:max. 0.005 %
- Nitrogen Compounds (as N):max. 0.001 %
- Residue after Ignition:max. 0.01 %
- Substances Darkened by Hot H2SO4:passes test
- Sulfate (SO4):max. 0.002 %
- Calcium (Ca):max. 5
- Chloride (Cl):max. 5
- Iron (Fe):max. 2
- Heavy Metals (as Pb):max. 5
Specification Test Results
About this item
:
:126.07 g/mol
Merck Index:13,06980
Boiling Pt:149…160 °C (1013 hPa)
Melting Pt:101 °C
Density:1,65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-634-3
UN:3261
ADR:8,III
REACH:01-2119534576-33
:GHS07
CAS Number:6153-56-6
Formula:HO₂CCO₂H·2H₂O
MDL Number:MFCD00149102