
Manganese(II) acetate tetrahydrate for synthesis, Sigma-Aldrich®
: Merck
:6156-78-1
:
:(H₃CCOO)₂Mn·4H₂O
:MFCD00062552
:245.09 g/mol
:13,05746
:300 °C
:1,58 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:211-334-3
:GHS08
:Ambient
- Solubility:330 g/l (20 °C)
- Bulk Density:700 kg/m3
- Density:1.59 g/cm3
- pH:7.0 (50 g/l, H2O, 20 °C)
- Hill Formula:C4H6MnO4*4H2O
- Chemical Formula:(CH3COO)2Mn*4H2O
Frequently Bought Together