Magnesium acetate tetrahydrate 98.0-102.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16674-78-5
:
Formula:(H₃CCOO)₂Mg·4H₂O
MDL Number:MFCD00149214
:214.46 g/mol
Merck Index:14,5654
Melting Pt:80 °C
Density:1,454
Storage Temperature:Ambient
EINECS:205-554-9
Related Information
Frequently Bought Together