Order Entry
Northern Ireland
ContactUsLinkComponent
Semicarbazide hydrochloride
Semicarbazide hydrochloride
Catalog # MOLEM83817869
Supplier:  Molekula
CAS Number:  
Semicarbazide hydrochloride
Catalog # MOLEM83817869
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :111.53 g/mol
Merck Index:12,08588
Melting Pt:174…178 °C
EINECS:209-247-0
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:563-41-7
Formula:CH₆ClN₃O
MDL Number:MFCD00013009