



Hydrazinium sulphate, GR for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:10034-93-2
:
Formula:NH₂NH₂·H₂SO₄
MDL Number:MFCD00044873
:130.12 g/mol
Merck Index:14,04772
Melting Pt:254 °C
Density:1,37 g/cm³ (20 °C)
EINECS:233-110-4
ADR:6.1,III
:M-Clarity™ quality level = MQ100
UN:3288
:GHS06
:GHS08
:GHS05
:GHS09
Specification Test Results
- Assay (iodometric):≥ 99.0%
- Identity:passes test
- Insoluble matter:≤0.005 %
- Chloride (Cl):≤0.0005 %
- Heavy metals (as Pb):≤0.0005 %
- As (Arsenic):≤ 0.0001 %
- Fe (Iron):≤ 0.001 %
- Residue on ignition (as SO.):≤ 0.05 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.04603.0100_TDS_EN ProductPage_104603
Product specification
msds_104603
SDS
Frequently Bought Together