
Guanidinium hydrochloride
Catalog # ROTH0035.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:95.53 g/mol
Merck Index:13,04578
Melting Pt:185 °C
Density:1,354 g/cm³ (20 °C)
EINECS:200-002-3
:GHS07
Storage Temperature:Ambient
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026