


4-(Methylamino)phenol sulphate
Catalog # MOLEM45828875
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:344.39 g/mol
Merck Index:13,06046
Boiling Pt:227 °C (1013 hPa)
Melting Pt:86 °C
Density:1,434 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-237-1
UN:2811
ADR:6.1,III
:GHS08
:GHS07
:GHS09
CAS Number:55-55-0
:51-72-9
Formula:C₁₄H₂₀N₂O₆S
MDL Number:MFCD00013140