Order Entry
Northern Ireland
ContactUsLinkComponent
N,N-Dimethyl-1,4-phenylenediammonium dichloride analytical reagent, Supelco®
Toxic
N,N-Dimethyl-1,4-phenylenediammonium dichloride analytical reagent, Supelco®
Catalog # 1.03067.0025
Supplier:  Merck
CAS Number:  
Toxic
N,N-Dimethyl-1,4-phenylenediammonium dichloride analytical reagent, Supelco®
Catalog # 1.03067.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    2000 g/l (20 °C)
  • Melting Point:
    208 - 212 °C (decomposition)
  • Molar Mass:
    209.11 g/mol
  • Bulk Density:
    500 kg/m3
  • pH:
    1.6 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

About this item

 :
 :209.12 g/mol
Merck Index:13,03280
Melting Pt:208…212 °C
Storage Temperature:Ambient
ADR:6.1,II
 :M-Clarity™ quality level = MQ100
EINECS:208-635-7
UN:2811
 :GHS06
CAS Number:536-46-9
 :MFCD00012990
Formula:(CH₃)₂NC₆H₄NH₂·2HCl
MDL Number:MFCD00012991