Order Entry
Northern Ireland
ContactUsLinkComponent
Mercury(II) sulphate 200 g/l in aqueous solution
Toxic
Health
Mercury(II) sulphate 200 g/l in aqueous solution
Catalog # MOLE61441891-1L
Supplier:  Molekula
CAS Number:  
Toxic
Health
Mercury(II) sulphate 200 g/l in aqueous solution
Catalog # MOLE61441891-1L
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :296.65 g/mol
EINECS:231-992-5
UN:2922
ADR:8,II
 :GHS06
 :GHS08
CAS Number:7783-35-9
Formula:HgSO₄
MDL Number:MFCD00011047