
Zinc sulphate 0.1 M ZnSO4 ≥99.5% volumetric solution, Fluka™
Catalog # 35392-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.999 - 1.001
Specification Test Results
About this item
:
ADR:9,III
:287.56 g/mol
EINECS:231-793-3
:GHS07
Storage Temperature:Ambient
CAS Number:7446-20-0
Formula:ZnSO₄·7H₂O
MDL Number:MFCD00011302