

Iodine standardised solution 1.0 N in ethanol
Catalog # 44488.K2
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
:253.81 g/mol
Storage Temperature:Ambient
EINECS:231-442-4
UN:2924
ADR:3,II
:GHS02
:GHS08