Sodium methoxide ∼30% (w/w) in methanol
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sodium methylate, Sodium methanolate
CAS Number:124-41-4
:
Formula:CH₃NaO
MDL Number:MFCD00012179
Boiling Pt:93 °C
Melting Pt:<6 °C
Density:0.969
Molecular Weight:54.02 g/mol
Flash Pt:16 °C (61 °F)
UN:1289
ADR:3 III
Merck Index:14,8643
EINECS:204-699-5
:GHS02
:GHS06
:GHS08
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together