Hydrochloric acid 0.1 M HCl (0.1N) 99.5-100.5% volumetric solution, Fluka™
318965-2L
:
- Pack type:Plastic bottle
- Pk:2 L
- Seq:5086
- Environmentally Preferable:
- Assay:0.0950 - 0.1050 N
- Traceable to NIST SRM:effective
- Vapor Pressure:190 mmHg ( 25 °C)
: Hydrochloric acid solution, Muriatic acid
:
:100 °C (1,013 hPa)
:1,000 g/cm³ (20 °C)
:Ambient
:1789
:8,III
:36.46 g/mol
:231-791-2
:GHS05
:7647-01-0
:HCl
:MFCD00011324