Order Entry
Northern Ireland
ContactUsLinkComponent
Glutaraldehyde 50% in aqueous solution, GPR RECTAPUR®
 
 
 
 
Glutaraldehyde 50% in aqueous solution, GPR RECTAPUR®
  VWRS28682HG
 :  
 
 
 
 
Glutaraldehyde 50% in aqueous solution, GPR RECTAPUR®
  VWRS28682HG
 :  

 

  • Pk:
    25 kg
  • Seq:
    4283

 

 

  • Assay:
    About 50 %
  • Methanol:
    Max. 0.5 %

 

 

 :
 :100.12 g/mol
 :1,13 g/cm³ (20 °C)
 :Refrigerator
 :203-856-5
 :2927
 :6.1,II
 :GHS06
 :GHS05
 :GHS08
 :GHS09
 :111-30-8
 :C₅H₈O₂
 :MFCD00007025