Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(±)-2-Methylpiperazine
(±)-2-Methylpiperazine
(±)-2-Methylpiperazine
MOLEM12067413
:
Molekula
:
(±)-2-Methylpiperazine
MOLEM12067413
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
100.16 g/mol
:
155 °C (1013 hPa)
:
Ambient
:
203-644-2
:
2925
:
4.1,II
:
109-07-9
:
C₅H₁₂N₂
:
MFCD00005954