Order Entry
Northern Ireland
ContactUsLinkComponent
 
Flammable
Irritant
(±)-2-Methyl-1-butanol for synthesis, Sigma-Aldrich®
Catalog # 8.06031.1000
Supplier:  Merck
CAS Number:  
Flammable
Irritant
(±)-2-Methyl-1-butanol for synthesis, Sigma-Aldrich®
Catalog # 8.06031.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    36 g/l (20 °C)
  • Melting Point:
    -70 °C
  • Molar Mass:
    88.15 g/mol
  • Boiling Point:
    127 - 129 °C
  • Vapor Pressure:
    4.3 hPa (20 °C)
  • Flash Point:
    42.5 °C
  • Refractive Index:
    1.4107 (20 °C, 589 nm)
  • Explosion Limit:
    1.2 - 10.3 %(V)
  • Density:
    0.82 g/cm3 (20 °C)
  • pH:
    7 (H2O)
  • Ignition Point:
    340 °C

Specification Test Results

About this item

 :
Merck Index:12,06109
Boiling Pt:129 °C (1013 hPa)
Melting Pt:–70 °C
Density:0,82 g/cm³ (20 °C)
ADR:3,III
 :M-Clarity™ quality level = MQ200
 :88.15 g/mol
Storage Temperature:Ambient
EINECS:205-289-9
UN:1993
 :GHS02
 :GHS07
CAS Number:137-32-6
Formula:C₅H₁₂O
MDL Number:MFCD00004743