Order Entry
Northern Ireland
ContactUsLinkComponent
(±)-2-Ethyl-1-hexanol for synthesis, Sigma-Aldrich®
Irritant
(±)-2-Ethyl-1-hexanol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Irritant
(±)-2-Ethyl-1-hexanol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    1.1 g/l (20 °C)
  • Melting Point:
    -76 °C
  • Molar Mass:
    130.23 g/mol
  • Boiling Point:
    184 °C (1013 hPa)
  • Vapor Pressure:
    0.13 hPa (20 °C)
  • Flash Point:
    75 °C DIN 51758
  • Refractive Index:
    1.4317 (20 °C, 589 nm)
  • Explosion Limit:
    1.1 - 12.7 %(V)
  • Density:
    0.832 g/cm3 (20 °C)
  • pH:
    7 (1 g/l, H2O, 20 °C)
  • Ignition Point:
    270 - 330 °C DIN 51794
  • Synonyms:
    Isocotyl alcohol, Isooctanol
  • Hill Formula:
    C8H18O

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together