Order Entry
Northern Ireland
ContactUsLinkComponent
DL-Leucine for synthesis, Sigma-Aldrich®
DL-Leucine for synthesis, Sigma-Aldrich®
Catalog # 8.14986.0050
Supplier:  Merck
CAS Number:  
DL-Leucine for synthesis, Sigma-Aldrich®
Catalog # 8.14986.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Solubility:
    7.97 g/l (0 °C)
  • Melting Point:
    290 - 295 °C
  • Molar Mass:
    131.17 g/mol
  • Density:
    1.293 g/cm3 (18 °C)

Specification Test Results

About this item

 :
 :131.17 g/mol
Merck Index:13,05470
Boiling Pt:226 °C (1013 hPa)
Density:1,191 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:206-328-2
CAS Number:328-39-2
Formula:C₆H₁₃NO₂
MDL Number:MFCD00063087