
3-Mercapto-1,2-propanediol for synthesis, Sigma-Aldrich®
: Merck
:96-27-5
:
:C₃H₈O₂S
:MFCD00004879
:108.16 g/mol
:12,09471
:118 °C (1013 hPa)
:1,247 g/cm³ (20 °C)
:202-495-0
:6.1,III
:M-Clarity™ quality level = MQ200
:2810
:GHS06
:Ambient
- Molar Mass:108.15 g/mol
- Boiling Point:118 °C (6.7 hPa)
- Flash Point:110 °C
- Density:1.247 g/cm3 (20 °C)
Frequently Bought Together