Order Entry
Northern Ireland
ContactUsLinkComponent
(±)-1,2-Hexanediol 98%
(±)-1,2-Hexanediol 98%
Catalog # ACRO200132500
CAS Number:  
undefined
(±)-1,2-Hexanediol 98%
Catalog # ACRO200132500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4410 to 1.4430 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :118.18 g/mol
Boiling Pt:223…224 °C (760 mmHg)
Melting Pt:–25 °C
Density:0,95 g/cm³
Flash Pt:122 °C
Storage Temperature:Ambient
EINECS:230-029-6
 :GHS07
CAS Number:6920-22-5
Formula:C₆H₁₄O₂
MDL Number:MFCD00010737