


Styrene ≥99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-42-5
:
Formula:C₈H₈
MDL Number:MFCD00008612
:104.15 g/mol
Merck Index:14,8860
Boiling Pt:145…146 °C
Melting Pt:–31 °C
Density:0,906
Flash Pt:31 °C (88 °F)
Storage Temperature:Refrigerator
UN:2055
ADR:3,III
:Stabilised with 4-tert-butylpyrocatechol (TBC) 10-15 ppm
EINECS:202-851-5
:GHS08
:GHS02
:GHS07
Related Information
Frequently Bought Together