Styrene stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:100-42-5
:
Formula:C₈H₈
MDL Number:MFCD00008612
:104.15 g/mol
Merck Index:13,08944
Boiling Pt:145,2 °C (1013 hPa)
Melting Pt:–31 °C
Density:0,906 g/cm³ (20 °C)
EINECS:202-851-5
UN:2055
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:31 °C
:GHS02
:GHS08
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:0.24 g/l (20 °C)
- Melting Point:-31 °C
- Molar Mass:104.15 g/mol
- Boiling Point:145 °C (1013 hPa)
- Vapor Pressure:6 hPa (20 °C)
- Flash Point:31 °C
- Refractive Index:1.5458 (20 °C, 589 nm)
- Explosion Limit:1.1 - 8.9 %(V)
- Density:0.906 g/cm3 (20 °C)
- Ignition Point:480 °C
- Saturation concentration:25.6 g/m3 (20 °C)
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Related Information
Frequently Bought Together