(±)-3-(Diethylamino)-1,2-propanediol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:621-56-7
:
Formula:C₇H₁₇NO₂
MDL Number:MFCD00004716
:147.22 g/mol
Boiling Pt:233…235 °C
Melting Pt:–30 °C
Density:0,970
Flash Pt:107 °C (224 °F)
Storage Temperature:Ambient
EINECS:210-693-3
ADR:8,II
:GHS05
Related Information
Frequently Bought Together