
DL-Malic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:617-48-1
Formula:HOOCCH(OH)CH₂COOH
MDL Number:MFCD00064212
:134.09 g/mol
Merck Index:13,05730
Boiling Pt:306 °C (1013 hPa)
Melting Pt:131…132 °C
Density:1,601 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:230-022-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:558 g/l (20 °C)
- Melting Point:128 - 132 °C
- Molar Mass:134.08 g/mol
- Bulk Density:800 kg/m3
- Flash Point:203 °C
- Density:1.60 g/cm3 (20 °C)
- pH:2.3 (10 g/l, H2O, 20 °C)
- Ignition Point:349 °C
Specification Test Results
Related Information
Frequently Bought Together