

Diethyl ether 99% stabilised for HPLC
Catalog # 38990.K2
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:74.12 g/mol
Merck Index:14,3806
Boiling Pt:34,6 °C
Melting Pt:–116 °C
Density:0,713
Flash Pt:–45 °C (–49 °F)
Storage Temperature:Ambient
EINECS:200-467-2
UN:1155
ADR:3,I
:Stabilised with ethanol
REACH:01-2119535785-29
:GHS02
:GHS07
CAS Number:60-29-7
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646