Order Entry
Northern Ireland
ContactUsLinkComponent
L(+)-Cysteine hydrochloride, Millipore®
L(+)-Cysteine hydrochloride, Millipore®
Catalog # 2430-1
CAS Number:  
L(+)-Cysteine hydrochloride, Millipore®
Catalog # 2430-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :157.62 g/mol
Merck Index:13,02810
Melting Pt:175 °C
Storage Temperature:Ambient
EINECS:200-157-7
CAS Number:52-89-1
Formula:HSCH₂CH(NH₂)COOH·HCl
MDL Number:MFCD00064553