Polyvinylpyrrolidone, M.W. ∼ 40,000, dry, TECHNICAL (K30)
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:9003-39-8
:
Formula:(C₆H₉NO)n
MDL Number:MFCD01076626
Storage Temperature:Ambient
Specification Test Results
- Identification:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number5L Plastic bucket UN Z (ZPACK13247005) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC26616.184 spec_VWRC26616.297
Product specification
Frequently Bought Together