Calcium-D-gluconate monohydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, E578, extra pure
Catalog # 1.02094.9029
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:448.39 g/mol
Melting Pt:178 °C
Storage Temperature:Ambient
EINECS:206-075-8
CAS Number:66905-23-5
:299-28-5
Formula:C₁₂H₂₂CaO₁₄·H₂O
MDL Number:MFCD00150787