
Polyethylene glycol 1,500 for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:25322-68-3
:
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839
:1500 g/mol
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:44…48 °C
Density:1,13 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:260 °C
EINECS:500-038-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:500 g/l (20 °C)
- Melting Point:44 - 48 °C
- Molar Mass:8000 g/mol
- Bulk Density:400 - 500 kg/m3
- Vapor Pressure:- 0.01 hPa (20 °C)
- Flash Point:260 °C
- Density:1.2 g/cm3 (20 °C)
- pH:4 - 7 (100 g/l, H2O, 20 °C)
- Ignition Point:420 °C
- Viscosity kinematic:33 - 39 mm2/s (20 °C) (50% solution)
Specification Test Results
Frequently Bought Together